Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016959.4 | NP_058655.3 | 2 | 216 | ||
GenBank mRNA | AK003158.1 | - | 2 | 142 | ||
AK010605.1 | - | 2 | 139 | |||
AK012345.1 | - | 2 | 139 | |||
AK050610.1 | - | 2 | 142 | |||
AK088113.1 | - | 2 | 139 | |||
AK135318.1 | - | 2 | 143 | |||
AK146304.1 | - | 2 | 150 | |||
AK148513.1 | - | 2 | 142 | |||
AK150441.1 | - | 2 | 142 | |||
AK150796.1 | - | 2 | 152 | |||
AK150814.1 | - | 2 | 142 | |||
AK151429.1 | - | 2 | 143 | |||
AK151982.1 | - | 2 | 152 | |||
AK153480.1 | - | 2 | 142 | |||
AK159777.1 | - | 2 | 142 | |||
AK167512.1 | - | 2 | 143 | |||
AK167562.1 | - | 2 | 141 | |||
AK168240.1 | - | 2 | 140 | |||
AK190515.1 | - | 2 | 113 | |||
AK196258.1 | - | 2 | 128 | |||
AK200497.1 | - | 2 | 113 | |||
AK207259.1 | - | 2 | 113 | |||
AK207595.1 | - | 2 | 105 | |||
AK207749.1 | - | 2 | 89 | |||
AK207971.1 | - | 2 | 113 | |||
AK208520.1 | - | 2 | 113 | |||
AL118161.1 | - | 2 | 221 | |||
BC039659.1 | - | 2 | 141 | 5025347 | ||
BC081451.1 | - | 2 | 129 | 6824188 | ||
BC083338.1 | - | 2 | 122 | 6820218 | ||
BC084675.1 | - | 2 | 133 | 5319512 | ||
BC106115.1 | - | 2 | 131 | 2599128 | ||
M88335.1 | - | 2 | 96 |
Gene Symbol: Sox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK189901.1 | - | 235 |