Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016979.1 | NP_058675.1 | 1 | 358 | ||
GenBank mRNA | AJ238004.1 | - | 1 | 241 | ||
AK034681.1 | - | 1 | 677 | |||
AK036432.1 | - | 1 | 323 | |||
AK037141.1 | - | 1 | 352 | |||
AK039088.1 | - | 1 | 373 | |||
AK081548.1 | - | 1 | 338 | |||
AK139510.1 | - | 1 | 366 | |||
AK149937.1 | - | 1 | 336 | |||
AK150588.1 | - | 1 | 320 | |||
AK152166.1 | - | 1 | 155 | |||
AK154447.1 | - | 1 | 338 | |||
AK169322.1 | - | 1 | 309 | |||
BC006875.1 | - | 1 | 358 | 3596013 |