Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016979.1 | NP_058675.1 | 3 | 753 | ||
GenBank mRNA | AJ238004.1 | - | 3 | 636 | ||
AK034681.1 | - | 3 | 1072 | |||
AK036432.1 | - | 3 | 718 | |||
AK037141.1 | - | 3 | 747 | |||
AK039088.1 | - | 3 | 768 | |||
AK081548.1 | - | 3 | 733 | |||
AK139510.1 | - | 3 | 761 | |||
AK149937.1 | - | 3 | 731 | |||
AK150588.1 | - | 3 | 715 | |||
AK152166.1 | - | 3 | 550 | |||
AK154447.1 | - | 3 | 733 | |||
AK169322.1 | - | 3 | 704 | |||
BC006875.1 | - | 3 | 753 | 3596013 |