Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnliprp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018874.2 | NP_061362.1 | 12 | 1430 | ||
GenBank mRNA | AF061274.1 | - | 12 | 1431 | ||
AK028105.1 | - | 12 | 1343 | |||
BC068266.1 | - | 8 | 986 | |||
BC090985.1 | - | 12 | 1342 | 30315884 |