Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019411.4 | NP_062284.1 | 3 | 626 | ||
GenBank mRNA | AF076192.1 | - | 3 | 596 | ||
AK076110.1 | - | 3 | 627 | |||
AK172644.1 | - | 3 | 599 | |||
AK195883.1 | - | 1 | 91 | |||
AK214382.1 | - | 1 | 19 | |||
BC003856.1 | - | 3 | 603 | 3484084 | ||
BC054458.1 | - | 3 | 586 | 6493866 | ||
Z67745.1 | - | 3 | 401 |