Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256672.1 | NP_001243601.1 | 5 | 878 | ||
NM_001256673.1 | NP_001243602.1 | 4 | 728 | |||
NM_001256674.1 | NP_001243603.1 | 4 | 728 | |||
NM_001256675.1 | NP_001243604.1 | 5 | 878 | |||
NM_001256676.1 | NP_001243605.1 | 5 | 692 | |||
NM_001256677.1 | NP_001243606.1 | 6 | 1009 | |||
NM_001324226.1 | NP_001311155.1 | 4 | 997 | |||
NM_001324227.1 | NP_001311156.1 | 5 | 692 | |||
NM_001324228.1 | NP_001311157.1 | 5 | 756 | |||
NM_001324229.1 | NP_001311158.1 | 4 | 1183 | |||
NM_001324230.1 | NP_001311159.1 | 6 | 1009 | |||
NM_004809.4 | NP_004800.2 | 5 | 878 | |||
XM_005254792.4 | XP_005254849.1 | 5 | 1782 | |||
XM_006720773.3 | XP_006720836.1 | 5 | 1783 | |||
GenBank mRNA | AF074953.1 | - | 5 | 743 | ||
AF111800.1 | - | 4 | 677 | |||
AF156557.1 | - | 5 | 886 | |||
AK075244.1 | - | 5 | 774 | |||
AK093689.1 | - | 1 | 1146 | |||
AK222665.1 | - | 5 | 761 | |||
AK300799.1 | - | 6 | 893 | |||
AK302051.1 | - | 4 | 683 | |||
AL109664.1 | - | 4 | 558 | |||
AL109665.1 | - | 5 | 732 | |||
AL533460.3 | - | 5 | 669 | |||
BC034379.1 | - | 5 | 824 | 5185908 | ||
BC065249.1 | - | 5 | 824 | 6059393 | ||
BX464941.2 | - | 5 | 692 | |||
CR542089.1 | - | 5 | 753 | |||
DQ064605.1 | - | 5 | 753 | |||
Y16522.1 | - | 5 | 795 |