Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019642.4 | NP_062616.2 | 7 | 1061 | ||
GenBank mRNA | AK004968.1 | - | 7 | 1062 | ||
AK018761.2 | - | 7 | 1100 | |||
AK087923.1 | - | 1 | 98 | |||
AK153952.1 | - | 7 | 922 | |||
AK161624.1 | - | 7 | 872 | |||
AK166778.1 | - | 7 | 847 | |||
AK167864.1 | - | 7 | 841 | |||
AK167866.1 | - | 7 | 841 | |||
AK168221.1 | - | 7 | 841 | |||
BC010509.1 | - | 6 | 834 | |||
BC046806.1 | - | 7 | 1089 | 6829209 | ||
D31717.1 | - | 6 | 847 |