Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019963.1 | NP_064347.1 | 2 | 264 | ||
GenBank mRNA | AF187231.1 | - | 2 | 258 | ||
AF206162.1 | - | 2 | 264 | |||
AK149838.1 | - | 2 | 261 | |||
AK149924.1 | - | 2 | 277 | |||
AK150070.1 | - | 2 | 276 |