Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019963.1 | NP_064347.1 | 20, 21 | 2019 | ||
GenBank mRNA | AF187231.1 | - | 20, 21 | 2016 | ||
AF206162.1 | - | 20, 21 | 2019 | |||
AK030903.1 | - | 13, 14 | 1111 | |||
AK149924.1 | - | 20, 21 | 2032 | |||
AK150070.1 | - | 20, 21 | 2031 | |||
AK153405.1 | - | 18, 19 | 1666 | |||
BC064827.1 | - | 9, 10 | 1016 |