Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004815.3 | NP_004806.3 | 2 | 515 | ||
XM_011542438.2 | XP_011540740.1 | 2 | 390 | |||
XM_011542439.2 | XP_011540741.1 | 2 | 235 | |||
GenBank mRNA | AB208873.1 | - | 1 | 121 | ||
AK307733.1 | - | 2 | 344 | |||
AK310491.1 | - | 2 | 235 | |||
BC022483.1 | - | 2 | 397 | |||
BC067839.1 | - | 2 | 510 | |||
BC093741.1 | - | 1 | 188 | 7939586 | ||
BC093767.1 | - | 1 | 191 | 7939612 | ||
BI548487.1 | - | 2 | 517 | |||
U90920.1 | - | 2 | 329 |