Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252679.1 | NP_001239608.1 | 5 | 376 | ||
NM_001252680.1 | NP_001239609.1 | 4 | 306 | |||
NM_001252681.1 | NP_001239610.1 | 3 | 239 | |||
NM_021289.2 | NP_067264.2 | 3 | 239 | |||
GenBank mRNA | BC111829.1 | - | 4 | 269 | 40055208 | |
U82376.1 | - | 4 | 306 | |||
U82377.1 | - | 3 | 239 | |||
U82378.1 | - | 3 | 239 | |||
U82379.1 | - | 5 | 376 |