Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077712.2 | NP_001071180.1 | 7 | 1018 | ||
NM_001290713.1 | NP_001277642.1 | 6 | 744 | |||
NM_021465.4 | NP_067440.3 | 7 | 878 | |||
GenBank mRNA | AJ002636.1 | - | 6 | 720 | ||
AK146718.1 | - | 7 | 878 | |||
AK168899.1 | - | 7 | 878 | |||
BC066041.1 | - | 7 | 759 | |||
CO434048.1 | - | 5 | 565 |