Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077712.2 | NP_001071180.1 | 19 | 2269 | ||
NM_001290713.1 | NP_001277642.1 | 18 | 1995 | |||
NM_021465.4 | NP_067440.3 | 19 | 2129 | |||
GenBank mRNA | AJ002636.1 | - | 18 | 1971 | ||
BC066041.1 | - | 19 | 2010 |