Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021880.2 | NP_068680.1 | 4 | 529 | ||
GenBank mRNA | AK005039.1 | - | 4 | 531 | ||
AK019147.1 | - | 2 | 164 | |||
AK027916.1 | - | 4 | 496 | |||
AK049832.1 | - | 4 | 522 | |||
AK051068.1 | - | 4 | 530 | |||
AK053918.1 | - | 4 | 530 | |||
AK083383.1 | - | 4 | 510 | |||
AK145797.1 | - | 4 | 530 | |||
AK145860.1 | - | 4 | 497 | |||
AK146380.1 | - | 4 | 516 | |||
AK147188.1 | - | 4 | 521 | |||
AK150427.1 | - | 4 | 522 | |||
AK150927.1 | - | 2 | 162 | |||
AK151040.1 | - | 2 | 162 | |||
AK151124.1 | - | 4 | 488 | |||
AK151556.1 | - | 2 | 162 | |||
AK152044.1 | - | 2 | 162 | |||
AK153227.1 | - | 4 | 488 | |||
AK158562.1 | - | 4 | 867 | |||
BC003461.1 | - | 4 | 500 | 2648357 | ||
BC005697.1 | - | 4 | 471 | 3597765 | ||
HQ412666.1 | - | 5 | 712 | |||
HQ412667.1 | - | 4 | 528 | |||
HQ412668.1 | - | 4 | 588 |