Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159516.1 | NP_001152988.1 | 2 | 675 | ||
NM_001159517.1 | NP_001152989.1 | 2 | 675 | |||
NM_021881.2 | NP_068681.1 | 2 | 675 | |||
GenBank mRNA | AF090402.1 | - | 3 | 835 | ||
AF090403.1 | - | 3 | 835 | |||
AF091047.1 | - | 835 | ||||
AK005169.1 | - | 2 | 61 | |||
AK132303.1 | - | 2 | 676 | |||
BC053426.1 | - | 3 | 347 | 6335004 | ||
BC056346.1 | - | 2 | 201 | 5705171 | ||
U44940.1 | - | 2 | 694 |