Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021882.4 | NP_068682.2 | 6 | 1167 | ||
GenBank mRNA | AF119092.1 | - | 1 | 44 | ||
AK012808.1 | - | 6 | 1168 | |||
AK131956.1 | - | 6 | 1169 | |||
AK178741.1 | - | 1 | 34 | |||
AK197730.1 | - | 1 | 34 | |||
BC075667.1 | - | 6 | 1152 | |||
BC082555.1 | - | 6 | 1153 | 30541702 | ||
U14133.1 | - | 6 | 1142 |