Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021882.4 | NP_068682.2 | 6 | 998 | ||
GenBank mRNA | AK012808.1 | - | 6 | 999 | ||
AK131956.1 | - | 6 | 1000 | |||
BC075667.1 | - | 6 | 983 | |||
BC082555.1 | - | 6 | 984 | 30541702 | ||
U14133.1 | - | 6 | 973 |