Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023133.1 | NP_075622.1 | 6 | 596 | ||
GenBank mRNA | AK012052.1 | - | 5 | 1253 | ||
AK013524.1 | - | 6 | 476 | |||
AK018725.1 | - | 5 | 455 | |||
AK028244.1 | - | 6 | 472 | |||
BC034506.1 | - | 6 | 467 | 3968247 | ||
BC086775.1 | - | 5 | 458 | |||
BC086938.1 | - | 6 | 473 | 30121701 |