Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163663.1 | NP_001157135.1 | 6 | 1121 | ||
NM_024287.4 | NP_077249.1 | 6 | 1121 | |||
GenBank mRNA | AB041575.1 | - | 6 | 920 | ||
AB232596.1 | - | 6 | 483 | |||
AK051246.1 | - | 6 | 956 | |||
AK083262.1 | - | 6 | 945 | |||
AK084131.1 | - | 6 | 929 | |||
AK154012.1 | - | 6 | 1018 | |||
AK160134.1 | - | 6 | 1000 | |||
AK160866.1 | - | 6 | 929 | |||
AK169842.1 | - | 6 | 1122 | |||
AK183477.1 | - | 5 | 404 | |||
AK185691.1 | - | 5 | 404 | |||
AK197621.1 | - | 2 | 123 | |||
BC019118.1 | - | 6 | 753 | 3711087 | ||
BC026915.1 | - | 5 | 411 |