Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089199.1 | - | 6 | 979 | ||
AK134691.1 | - | 6 | 865 | |||
AK159832.1 | - | 6 | 914 | |||
AK167599.1 | - | 6 | 914 | |||
AK170540.1 | - | 6 | 934 | |||
AK172328.1 | - | 6 | 871 | |||
BC027378.1 | - | 5 | 686 | |||
BC029678.1 | - | 6 | 864 | 5368061 |