Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025285.2 | NP_079561.1 | 3, 4 | 369 | ||
GenBank mRNA | AK003659.1 | - | 3, 4 | 369 | ||
AK013020.1 | - | 3, 4 | 370 | |||
AK149212.1 | - | 3, 4 | 394 | |||
BC026538.1 | - | 3, 4 | 331 | 4981703 |