Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ren1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031192.3 | NP_112469.1 | 1, 2 | 117 | ||
GenBank mRNA | AK052685.1 | - | 1, 2 | 117 | ||
AK085309.1 | - | 1, 2 | 117 | |||
BC024331.1 | - | 1, 2 | 88 | |||
BC061053.1 | - | 1, 2 | 115 | 30313659 | ||
X16642.1 | - | 1, 2 | 99 |