Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 2 | 260 | ||
NM_031196.3 | NP_112473.1 | 2 | 354 | |||
GenBank mRNA | AK002560.1 | - | 2 | 271 | ||
AK050714.1 | - | 2 | 384 | |||
AK079592.1 | - | 2 | 315 | |||
AK085024.2 | - | 2 | 300 | |||
AK088803.1 | - | 2 | 317 | |||
AK138577.1 | - | 2 | 315 | |||
BC015263.1 | - | 2 | 272 | 4239690 | ||
BF786711.1 | - | 2 | 271 | |||
L23755.1 | - | 1 | 212 | |||
L36539.1 | - | 2 | 267 | |||
U32469.1 | - | 2 | 294 | |||
U66103.1 | - | 2 | 307 |