Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 6 | 1520 | ||
NM_031196.3 | NP_112473.1 | 6 | 1614 | |||
GenBank mRNA | AK002560.1 | - | 6 | 1531 | ||
AK079592.1 | - | 6 | 1575 | |||
AK085024.2 | - | 6 | 1562 | |||
AK088803.1 | - | 6 | 1577 | |||
AK138577.1 | - | 6 | 1440 | |||
BC015263.1 | - | 6 | 1532 | 4239690 | ||
L23755.1 | - | 5 | 1472 | |||
L36539.1 | - | 6 | 1527 | |||
U32469.1 | - | 6 | 1554 | |||
U66103.1 | - | 6 | 1567 |