Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 6 | 1845 | ||
NM_031196.3 | NP_112473.1 | 6 | 1939 | |||
GenBank mRNA | AK002560.1 | - | 6 | 1856 | ||
AK079592.1 | - | 6 | 1900 | |||
AK088803.1 | - | 6 | 1902 | |||
AK138577.1 | - | 6 | 1765 | |||
BC015263.1 | - | 6 | 1857 | 4239690 | ||
L23755.1 | - | 5 | 1797 | |||
L36539.1 | - | 6 | 1852 | |||
U32469.1 | - | 6 | 1879 | |||
U66103.1 | - | 6 | 1892 |