Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004833.1 | NP_004824.1 | 2, 3 | 499 | ||
XM_017002848.1 | XP_016858337.1 | 3, 4 | 1726 | |||
GenBank mRNA | AF024714.1 | - | 2, 3 | 499 | ||
AK292042.1 | - | 2, 3 | 408 | |||
BC010940.1 | - | 2, 3 | 501 | 4080360 |