Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252332.1 | NP_001239261.1 | 4 | 635 | ||
NM_001252333.1 | NP_001239262.1 | 3, 4 | 629 | |||
NM_033144.2 | NP_149156.1 | 4 | 635 | |||
GenBank mRNA | AB093215.1 | - | 4 | 485 | ||
AF179996.1 | - | 3 | 331 | |||
AK144178.1 | - | 3, 4 | 471 | |||
BC037707.1 | - | 4 | 423 | |||
BC049819.1 | - | 4 | 450 | |||
BC059248.1 | - | 4 | 571 | 6400429 |