Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 3, 4 | 392 | ||
NM_033475.3 | NP_258436.2 | 2, 3 | 737 | |||
GenBank mRNA | AB082927.1 | - | 2, 3 | 650 | ||
AB232630.1 | - | 2, 3 | 148 | |||
AF322068.1 | - | 2, 3 | 148 | |||
AF327929.1 | - | 3, 4 | 342 | |||
AK160308.1 | - | 706 | ||||
AK200345.1 | - | 1, 2 | 68 | |||
AK205613.1 | - | 1, 2 | 19 | |||
AK205647.1 | - | 1, 2 | 19 | |||
AK216992.1 | - | 1, 2 | 19 | |||
BC038638.1 | - | 2, 3 | 212 | 5058535 |