Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 11 | 974 | ||
NM_033475.3 | NP_258436.2 | 10 | 1319 | |||
GenBank mRNA | AB082927.1 | - | 10 | 1232 | ||
AB232630.1 | - | 10 | 730 | |||
AF322068.1 | - | 10 | 730 | |||
AF327929.1 | - | 11 | 924 | |||
AK160308.1 | - | 1288 | ||||
AK181473.1 | - | 3 | 144 | |||
BC038638.1 | - | 10 | 794 | 5058535 | ||
S72304.1 | - | 8 | 577 |