Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 5 | 479 | ||
NM_033475.3 | NP_258436.2 | 4 | 824 | |||
GenBank mRNA | AB082927.1 | - | 4 | 737 | ||
AB232630.1 | - | 4 | 235 | |||
AF322068.1 | - | 4 | 235 | |||
AF327929.1 | - | 5 | 429 | |||
AK160308.1 | - | 793 | ||||
AK179020.1 | - | 2 | 107 | |||
AK200345.1 | - | 3 | 155 | |||
AK205613.1 | - | 3 | 106 | |||
AK205647.1 | - | 3 | 106 | |||
AK216992.1 | - | 3 | 106 | |||
BC038638.1 | - | 4 | 299 | 5058535 | ||
S72304.1 | - | 2 | 82 |