Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146260.2 | NP_666372.1 | 5 | 1463 | ||
GenBank mRNA | AF481143.1 | - | 5 | 1463 | ||
AK030341.1 | - | 5 | 1493 | |||
AK031966.1 | - | 5 | 1525 | |||
BC138840.1 | - | 5 | 1027 | 8861862 | ||
BC141124.1 | - | 5 | 1027 | 9055736 |