Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162365.1 | NP_001155837.1 | 25, 26 | 2705 | ||
NM_001162366.1 | NP_001155838.1 | 24, 25 | 2535 | |||
NM_172498.3 | NP_766086.2 | 24, 25 | 2579 | |||
GenBank mRNA | AA462640.1 | - | 3, 4 | 253 | ||
AI181574.1 | - | 3, 4 | 319 | |||
AK088696.1 | - | 24, 25 | 2580 | |||
AK150105.1 | - | 25, 26 | 2636 | |||
BC024594.1 | - | 5, 6 | 581 | |||
BC137704.1 | - | 24, 25 | 2530 | 8860723 | ||
BC144849.1 | - | 24, 25 | 2530 | 9053397 | ||
JN971017.1 | - | 24, 25 | 2498 |