Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205367.1 | NP_001192296.1 | 13 | 2058 | ||
NM_009859.4 | NP_033989.2 | 14 | 2061 | |||
GenBank mRNA | AK088997.1 | - | 14 | 1848 | ||
AK144127.1 | - | 14 | 1852 | |||
AK220447.1 | - | 12 | 4104 | |||
BC058587.1 | - | 13 | 1862 | 6529169 |