Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023434.3 | NP_075923.2 | 4 | 450 | ||
GenBank mRNA | AF228408.1 | - | 2 | 109 | ||
AK041829.1 | - | 3 | 187 | |||
AK042459.1 | - | 3 | 187 | |||
AK088144.1 | - | 3 | 122 | |||
AK147023.1 | - | 3 | 156 | |||
AK147834.1 | - | 3 | 127 | |||
AK153278.1 | - | 3 | 116 | |||
AK173012.1 | - | 3 | 154 | |||
BC050091.1 | - | 2 | 105 | 30023547 |
Gene Symbol: Itgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC080751.1 | - | 104 |