Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023434.3 | NP_075923.2 | 10 | 2603 | ||
GenBank mRNA | AF228408.1 | - | 8 | 2262 | ||
AK007946.1 | - | 1 | 76 | |||
AK019980.1 | - | 6 | 1791 | |||
AK088144.1 | - | 9 | 2276 | |||
AK137050.1 | - | 828 | ||||
AK147023.1 | - | 9 | 2309 | |||
AK147834.1 | - | 9 | 2280 | |||
AK153278.1 | - | 9 | 2269 | |||
AK173012.1 | - | 9 | 2307 | |||
BC020135.1 | - | 1 | 215 | |||
BC050091.1 | - | 8 | 2259 | 30023547 |