Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286002.1 | NP_001272931.1 | 13 | 1744 | ||
NM_001286003.1 | NP_001272932.1 | 13 | 1995 | |||
NM_028776.5 | NP_083052.1 | 13 | 1970 | |||
GenBank mRNA | AK004809.2 | - | 13 | 1710 | ||
AK030824.1 | - | 13 | 1945 | |||
AK044100.1 | - | 13 | 1956 | |||
AK047267.1 | - | 1 | 751 | |||
AK162937.1 | - | 13 | 1745 | |||
BC043085.1 | - | 13 | 1671 | 6417440 | ||
BC066800.1 | - | 13 | 1655 |