Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029926.5 | NP_084202.2 | 12 | 1836 | ||
GenBank mRNA | AK028837.1 | - | 12 | 1776 | ||
AK029028.1 | - | 12 | 1832 | |||
AK078803.1 | - | 2 | 425 | |||
BC051676.1 | - | 12 | 1830 | 30017484 |