Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160420.2 | NP_001153892.1 | 4 | 521 | ||
NM_001160421.2 | NP_001153893.1 | 3 | 632 | |||
NM_001160422.1 | NP_001153894.1 | 3 | 632 | |||
NM_054076.3 | NP_473417.2 | 3 | 632 | |||
GenBank mRNA | AF333980.1 | - | 3 | 631 | ||
AK053318.1 | - | 3 | 415 | |||
AK153750.1 | - | 4 | 522 | |||
AY077682.1 | - | 3 | 624 | |||
BC099435.1 | - | 3 | 465 | 30788813 | ||
BC115928.1 | - | 3 | 585 | 40104738 |