Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080850.2 | NP_543126.2 | 3 | 588 | ||
GenBank mRNA | AK028435.1 | - | 3 | 590 | ||
AK083528.1 | - | 3 | 559 | |||
AK083590.1 | - | 3 | 553 | |||
AK129063.1 | - | 3 | 478 | |||
BC150821.1 | - | 2 | 300 | 9087732 |