Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139228.3 | NP_631974.1 | 3 | 464 | ||
GenBank mRNA | AJ313479.1 | - | 3 | 464 | ||
AK041678.1 | - | 1 | 103 | |||
BC054784.1 | - | 3 | 300 | 6817213 | ||
BC056969.1 | - | 3 | 300 | 6817213 |