Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139309.4 | NP_647470.1 | 4 | 545 | ||
GenBank mRNA | AB077383.1 | - | 5 | 729 | ||
AJ511807.1 | - | 5 | 565 | |||
AK052357.1 | - | 6 | 703 | |||
AK085931.1 | - | 4 | 546 | |||
AK142744.1 | - | 705 | ||||
AK144743.1 | - | 5 | 574 | |||
BC017538.1 | - | 5 | 561 | 4506804 |