Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139310.1 | NP_647471.1 | 4, 5 | 225 | ||
GenBank mRNA | AK157904.1 | - | 4, 5 | 257 | ||
AY055122.1 | - | 4, 5 | 225 |