Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139311.2 | NP_647472.2 | 10 | 1402 | ||
GenBank mRNA | AK143477.1 | - | 6 | 845 | ||
AY050217.1 | - | 10 | 1549 |