Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bivm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144558.4 | NP_653141.3 | 4 | 1055 | ||
GenBank mRNA | AF411386.1 | - | 5 | 1058 | ||
AK032375.1 | - | 4 | 1015 | |||
AK034329.1 | - | 4 | 1017 | |||
BC082584.1 | - | 4 | 861 | 30612339 |