Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144559.2 | NP_653142.2 | 4 | 579 | ||
GenBank mRNA | AF499613.1 | - | 4 | 547 | ||
AK076532.1 | - | 4 | 585 | |||
AK087863.1 | - | 4 | 579 | |||
AK170920.1 | - | 4 | 551 | |||
BC027310.1 | - | 4 | 566 | 3674499 | ||
EU050648.1 | - | 4 | 525 |