Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144933.1 | NP_659182.1 | 3, 4 | 699 | ||
GenBank mRNA | AK033151.1 | - | 3, 4 | 705 | ||
AK038451.1 | - | 3, 4 | 739 | |||
AK049112.1 | - | 3, 4 | 739 | |||
AK142033.1 | - | 3, 4 | 738 | |||
AK151377.1 | - | 3, 4 | 739 | |||
BC004062.1 | - | 2, 3 | 376 | |||
BC020185.1 | - | 3, 4 | 699 | 4505959 |