Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031141.1 | - | 3, 4 | 389 | ||
AK161676.1 | - | 3, 4 | 446 | |||
BC017156.1 | - | 3, 4 | 510 | 4911201 |