Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144941.3 | NP_659190.3 | 6 | 1054 | ||
GenBank mRNA | AK122462.1 | - | 1 | 940 | ||
AK208379.1 | - | 1 | 62 | |||
BC023677.1 | - | 6 | 1029 | 5354497 | ||
BM944941.1 | - | 4 | 345 | |||
CV563343.1 | - | 5 | 625 |