Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145099.2 | NP_659567.2 | 7, 8 | 844 | ||
GenBank mRNA | AB083654.1 | - | 7, 8 | 820 | ||
AF510316.1 | - | 7, 8 | 844 | |||
BC108984.1 | - | 7, 8 | 830 | 40047664 | ||
BC108985.1 | - | 7, 8 | 830 |