Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145156.1 | NP_660138.1 | 4 | 1341 | ||
GenBank mRNA | AF377993.1 | - | 3 | 810 | ||
AF377994.1 | - | 4 | 1159 | |||
AK008810.1 | - | 1 | 38 | |||
AK034173.1 | - | 1 | 2181 | |||
AK155044.1 | - | 2 | 3534 | |||
BC023172.1 | - | 2 | 852 | |||
BC025908.1 | - | 4 | 1341 | 5043992 |